Learn More
Grainger V-Belt Pulley Cast Iron 5.95 O.D.
Catalog No. 502748947
Encompass_Preferred
The V-Belt Pulley BK62-145U49 from Grainger is a V-belt pulley made of cast iron with a 1-7/16" bore. It has a 4-1/2" outside diameter and a 3/8" face width. This pulley is designed for use with fractional horsepower motors and can be used to change the speed ratio between a driving and driven shaft. It features a crowned face and is balanced for smooth operation. The cast iron material provides durability and resistance to wear. This is a high quality pulley well-suited for industrial power transmission applications.
Catalog No. 50-274-8947 Supplier Grainger Supplier No. 145U49
The V-Belt Pulley BK62-145U49 from Grainger is a V-belt pulley made of cast iron with a 1-7/16" bore. It has a 4-1/2" outside diameter and a 3/8" face width. This pulley is designed for use with fractional horsepower motors and can be used to change the speed ratio between a driving and driven shaft. It features a crowned face and is balanced for smooth operation. The cast iron material provides durability and resistance to wear. This is a high quality pulley well-suited for industrial power transmission applications.