Learn More
Grainger V-Belt Pulley Cast Iron 5.95 O.D.
Catalog No. 502752353
Encompass_Preferred
The V-Belt Pulley Fixed Bore 145R96 is a fixed bore pulley made by Browning. It has a 3.00 inch outside diameter, 1.50 inch bore diameter, and 0.50 inch face width. The pulley is made from cast iron with a rough finish and is designed for use with v-belts. It features set screws for securing to a shaft. This pulley can be used for power transmission applications.
Catalog No. 50-275-2353 Supplier Grainger Supplier No. 145R96
The V-Belt Pulley Fixed Bore 145R96 is a fixed bore pulley made by Browning. It has a 3.00 inch outside diameter, 1.50 inch bore diameter, and 0.50 inch face width. The pulley is made from cast iron with a rough finish and is designed for use with v-belts. It features set screws for securing to a shaft. This pulley can be used for power transmission applications.