Learn More
Grainger V-Belt Pulley Cast Iron 6.45 O.D.
Catalog No. 502749213
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 1-1/2" bore diameter and is made of cast iron. The pulley has a 4L cross section and is 6" in diameter. It is used with fractional horsepower motors and can be mounted on a motor shaft or jackshaft. This pulley allows power transmission from motors to drive mechanical equipment using a V-belt. It is a high quality, durable product made in the USA.
Catalog No. 50-274-9213 Supplier Grainger Supplier No. 145U53
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 1-1/2" bore diameter and is made of cast iron. The pulley has a 4L cross section and is 6" in diameter. It is used with fractional horsepower motors and can be mounted on a motor shaft or jackshaft. This pulley allows power transmission from motors to drive mechanical equipment using a V-belt. It is a high quality, durable product made in the USA.