Learn More
Grainger V-Belt Pulley Cast Iron 6.45 O.D.
Catalog No. 502748948
Encompass_Preferred
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley for use with v-belts. It has a cast iron construction with a painted finish. The pulley has a fixed 1-7/16 inch bore and is available in multiple sizes ranging from 2-1/2 to 12 inch outer diameters. The face widths also vary based on outer diameter size. This heavy duty pulley can be used for power transmission applications and with v-belt drives. It's designed for industrial and commercial settings.
Catalog No. 50-274-8948 Supplier Grainger Supplier No. 145U55
The V-Belt Pulley Fixed Bore from Grainger is a fixed bore pulley for use with v-belts. It has a cast iron construction with a painted finish. The pulley has a fixed 1-7/16 inch bore and is available in multiple sizes ranging from 2-1/2 to 12 inch outer diameters. The face widths also vary based on outer diameter size. This heavy duty pulley can be used for power transmission applications and with v-belt drives. It's designed for industrial and commercial settings.