Learn More
Grainger V-Belt Pulley Cast Iron 6.95 O.D.
Catalog No. 502753443
Encompass_Preferred
The V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley made by Browning. It has a 4.5 inch outside diameter, 1.25 inch bore diameter, and 0.5 inch face width. The pulley is made from cast iron with a black oxide finish. It is designed for use with v-belts and has a crowned face to keep the v-belt centered. The pulley can be mounted on a shaft with a locking collar or set screw. It is commonly used in industrial machinery belt drive systems. This heavy duty pulley provides efficient power transmission.
Catalog No. 50-275-3443 Supplier Grainger Supplier No. 145T09
The V-Belt Pulley Fixed Bore 145T09 is a fixed bore pulley made by Browning. It has a 4.5 inch outside diameter, 1.25 inch bore diameter, and 0.5 inch face width. The pulley is made from cast iron with a black oxide finish. It is designed for use with v-belts and has a crowned face to keep the v-belt centered. The pulley can be mounted on a shaft with a locking collar or set screw. It is commonly used in industrial machinery belt drive systems. This heavy duty pulley provides efficient power transmission.