Learn More
Grainger V-Belt Pulley Cast Iron 6.95 O.D.
Catalog No. 502752320
Encompass_Preferred
The Grainger 145T08 V-Belt Pulley is a fixed bore pulley with a 3.25" outside diameter, 1.75" bore diameter, and 0.5" face width. It has a cast iron construction and is used with v-belts for power transmission applications. The pulley has a set screw for securing it to a shaft. It is rated for speeds up to 3600 RPM. The pulley weighs approximately 2 lbs and has a black finish. It is made in the USA by Browning Manufacturing.
Catalog No. 50-275-2320 Supplier Grainger Supplier No. 145T08
The Grainger 145T08 V-Belt Pulley is a fixed bore pulley with a 3.25" outside diameter, 1.75" bore diameter, and 0.5" face width. It has a cast iron construction and is used with v-belts for power transmission applications. The pulley has a set screw for securing it to a shaft. It is rated for speeds up to 3600 RPM. The pulley weighs approximately 2 lbs and has a black finish. It is made in the USA by Browning Manufacturing.