Learn More
Grainger V-Belt Pulley Cast Iron 7.25 O.D.
Catalog No. 502749155
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial V-belt drives and compatible with type A, B, C, D, and E V-belts. The maximum bore RPM is 3600. This is a high quality, durable V-belt pulley for industrial applications.
Catalog No. 50-274-9155 Supplier Grainger Supplier No. 145P27
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 3.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial V-belt drives and compatible with type A, B, C, D, and E V-belts. The maximum bore RPM is 3600. This is a high quality, durable V-belt pulley for industrial applications.