Learn More
Grainger V-Belt Pulley Cast Iron 7.75 O.D.
Catalog No. 502749076
Encompass_Preferred
The Grainger 145U79 is a Westward 145U79 1/4-Inch Drive Click Torque Wrench that delivers between 10 and 50 inch pounds of torque. It features a knurled grip handle and a reversible ratchet head. The wrench has legible, high contrast markings and numbers on the handle to clearly indicate the torque setting. It can be adjusted by rotating the grip to the desired torque level. The wrench is made of chrome vanadium steel for durability. It weighs 0.60 lbs and is 12 inches in overall length.
Catalog No. 50-274-9076 Supplier Grainger Supplier No. 145U79
The Grainger 145U79 is a Westward 145U79 1/4-Inch Drive Click Torque Wrench that delivers between 10 and 50 inch pounds of torque. It features a knurled grip handle and a reversible ratchet head. The wrench has legible, high contrast markings and numbers on the handle to clearly indicate the torque setting. It can be adjusted by rotating the grip to the desired torque level. The wrench is made of chrome vanadium steel for durability. It weighs 0.60 lbs and is 12 inches in overall length.