Learn More
Grainger V-Belt Pulley Cast Iron 8.25 O.D.
Catalog No. 502753250
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 1-3/8" bore diameter and is made of cast iron for durability. The pulley has a 4L cross section and is 6" in diameter. It is designed for use with fractional horsepower motors in light-duty tension applications. The maximum RPM rating is 3600. This pulley allows for misalignment control in V-belt drive systems. It comes with a pre-lubricated sleeve bushing installed in the bore.
Catalog No. 50-275-3250 Supplier Grainger Supplier No. 145P32
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 1-3/8" bore diameter and is made of cast iron for durability. The pulley has a 4L cross section and is 6" in diameter. It is designed for use with fractional horsepower motors in light-duty tension applications. The maximum RPM rating is 3600. This pulley allows for misalignment control in V-belt drive systems. It comes with a pre-lubricated sleeve bushing installed in the bore.