Learn More
Grainger V-Belt Pulley Cast Iron 8.25 O.D.
Catalog No. 502749033
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing in the bore to accommodate a shaft. It has a cast iron hub and is made of cast iron for durability. The pulley has a QD bushing with set screws for easy installation and secure mounting. It comes in sizes from 2" to 12" outer diameter, with bores from 5/8" to 1-7/8". This heavy duty pulley is well-suited for industrial applications requiring power transmission via v-belts.
Catalog No. 50-274-9033 Supplier Grainger Supplier No. 145P31
The V-Belt Pulley Bushed Bore from Baldor is a V-belt pulley with a bushing in the bore to accommodate a shaft. It has a cast iron hub and is made of cast iron for durability. The pulley has a QD bushing with set screws for easy installation and secure mounting. It comes in sizes from 2" to 12" outer diameter, with bores from 5/8" to 1-7/8". This heavy duty pulley is well-suited for industrial applications requiring power transmission via v-belts.