Learn More
Grainger V-Belt Pulley Cast Iron 8.75 O.D.
Catalog No. 502753474
Encompass_Preferred
The Grainger 145U89 is a Westward 145U89 1/4-Inch Drive Click Torque Wrench that delivers between 10 and 50 inch pounds of torque. It features a knurled grip handle for comfort and control. The wrench has a reversible ratchet head that allows for tightening and loosening fasteners. It also has a positive lock with torque adjustment and a quick release button to easily remove sockets. The 145U89 is made in the USA and comes with a 1 year warranty from Westward. It is an accurate and durable torque wrench for precise tightening applications.
Catalog No. 50-275-3474 Supplier Grainger Supplier No. 145U89
The Grainger 145U89 is a Westward 145U89 1/4-Inch Drive Click Torque Wrench that delivers between 10 and 50 inch pounds of torque. It features a knurled grip handle for comfort and control. The wrench has a reversible ratchet head that allows for tightening and loosening fasteners. It also has a positive lock with torque adjustment and a quick release button to easily remove sockets. The 145U89 is made in the USA and comes with a 1 year warranty from Westward. It is an accurate and durable torque wrench for precise tightening applications.