Learn More
Grainger V-Belt Pulley Cast Iron 9.25 O.D.
Catalog No. 502753251
Encompass_Preferred
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 2.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. This is a high quality, durable V-belt pulley well-suited for industrial use.
Catalog No. 50-275-3251 Supplier Grainger Supplier No. 145P35
The V-Belt Pulley Bushed Bore from Grainger is a V-belt pulley with a bushing bore. It has a 2.25 inch outside diameter, a 1.25 inch bore, and a 0.5 inch bushing. The pulley is made from cast iron with an oil-impregnated bronze bushing. It is designed for industrial power transmission applications using a V-belt. This is a high quality, durable V-belt pulley well-suited for industrial use.