Learn More
MSC MSA Chin Protector, Retractable, Black, 1 ea
Catalog No. 5000788056
Encompass
V-Gard chin protectors work as a system with select V-Gard visors (nitrometers) to provide extra protection from impact and splash hazards to the face and neck. This retractable chin protector offers protection from impact and splash hazards to the face and neck area, making it suitable for various industrial applications.
  • Protection from impact and splash hazards
  • Retractable design with flexible plates for greater head movement flexibility
  • Approved for use with special versions of V-Gard visors (Nitrometers)
  • Opaque material for safety and compliance with various V-Gard visors
Catalog No. 50-007-88056 Supplier MSC Supplier No. 66646266
V-Gard chin protectors work as a system with select V-Gard visors (nitrometers) to provide extra protection from impact and splash hazards to the face and neck. This retractable chin protector offers protection from impact and splash hazards to the face and neck area, making it suitable for various industrial applications.
  • Protection from impact and splash hazards
  • Retractable design with flexible plates for greater head movement flexibility
  • Approved for use with special versions of V-Gard visors (Nitrometers)
  • Opaque material for safety and compliance with various V-Gard visors