Learn More
Jorgensen Labs Advanced Supraglottic Airway Device, Rabbit, R3
SDP
Catalog No. NC1845017
Encompass_Preferred
Widely used in human medicine, a supraglottic airway device (SGAD) is an alternative to an endotracheal tube, used to deliver anaesthetic gases and/or oxygen to anaesthetised patients. Designed to sit in the pharynx, they avoid many of the problems caused by ET tubes such as laryngeal and tracheal trauma. They are also quick and easy to place and are useful for emergency resuscitation. v-gel devices are the first veterinary species-specific SGADs, currently used all over the world in cats and rabbits.
Catalog No. NC1845017 Supplier Jorgensen Labs Supplier No. J1350CN
Widely used in human medicine, a supraglottic airway device (SGAD) is an alternative to an endotracheal tube, used to deliver anaesthetic gases and/or oxygen to anaesthetised patients. Designed to sit in the pharynx, they avoid many of the problems caused by ET tubes such as laryngeal and tracheal trauma. They are also quick and easy to place and are useful for emergency resuscitation. v-gel devices are the first veterinary species-specific SGADs, currently used all over the world in cats and rabbits.