Learn More
Fisherbrand™ V-Maxx™ Chemical Splash and Impact Goggles
SCI_ED
Catalog No. S68660 Shop All Fisherbrand Products
Change view
Click to view available options
Impact Goggle; Neoprene headband
Catalog No.
S68660 Impact Goggle; Neoprene headband
1 options
This item is not returnable. View return policy
Catalog No. S68660
This item is not returnable. View return policy
Comfortable fit ensures protection — even for contact wearers.
Offer protection against impact, dust, mist and chemical splash.
  • FogBan™ lens coating
  • Wraparound design provides maximum coverage and 180° field of vision
  • Soft rubber construction ensures secure, comfortable fit
  • Large enough to be worn over prescription glasses
Compliance
Meet ANSI™ Z87.1-2003 and CSA™ Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2003 and CSA Z94.3 standards
Lens Tint Clear
Product Type Impact Goggle
Ventilation Details Indirect
Material Polycarbonate