Learn More
PDC V-Notch™ Cerner Direct Thermal Paper Label, 250 Labels/Roll, 8 Rolls/Box
Catalog No. 21235834
Change view
Click to view available options
Quantity:
250 Labels/Roll, 8 Rolls/Box
Catalog No. Quantity
21-235-834 250 Labels/Roll, 8 Rolls/Box
1 options
Catalog No. 21-235-834 Supplier PDC Supplier No. TD751142BD
  • $89.00
    $66.65 / Case of 8
    Save $22.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Secondary labels for Cerner systems designed for quick, reliable identification of BD Vacutainer™ Plus Plastic Tubes with precise alignment.
  • Permanent adhesive for dependable use
  • Unique V-shaped notch ensures precise alignment on BD Vacutainer™ Plus Plastic Tubes
  • Black sensor bar supports accurate, jam-free printing and operational efficiency
  • Enhances patient safety by preventing misidentification and re-draws
  • Open window allows sample visibility for confirmation of proper fill
  • Compatible with Cerner™ information systems to streamline lab workflows
  • Ideal for healthcare and laboratory applications requiring reliable, high-clarity labeling
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 250 Labels/Roll, 8 Rolls/Box
Dimensions (L x W) 2 x 1 1/4 in.
Length (English) 2 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm