Learn More
PDC V-Notch™ Direct Thermal Label, 200/Roll, 8 Rolls/Box
Catalog No. 21235836
Change view
Click to view available options
Quantity:
200/Roll, 8 Rolls/Box
Catalog No. Quantity
21-235-836 200/Roll, 8 Rolls/Box
1 options
Catalog No. 21-235-836 Supplier PDC Supplier No. TD7523162BD
Add to Cart
Add to Cart
Secondary labels provide fast, reliable ID for BD Vacutainer™ Plus Tubes, enhancing patient safety and barcode readability.
  • Permanent adhesive for dependable performance
  • V-Notch™ design ensures precise alignment on BD Vacutainer™ Plus Plastic Tubes
  • Enhances barcode scanning accuracy and reduces labeling errors
  • Supports efficient workflow for nurses, phlebotomists, and lab technicians
  • Helps ensure proper tube fill and minimizes patient redraws
  • Streamlines laboratory operations while reducing costs and workflow interruptions
  • Ideal for healthcare facilities requiring reliable and consistent sample identification
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 200/Roll, 8 Rolls/Box
Dimensions (L x W) 2 x 2.18 in.
Length (English) 2 in.
Width (English) 2.1875 in.
Width (Metric) 55.5625 mm