Learn More
Meso Scale Discovery V-PLEX Ab38 Peptide 4G8 Kit
SDP
Catalog No. 5000078321
Encompass
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody calibrator and assay diluent optimized for A 38 detection With a lower limit of detection of 1 95 pg/mL this kit enables the accurate quantification of A 38 for research into Alzheimer s disease and other neurodegenerative disorders
Catalog No. 50-000-78321 Supplier Meso Scale Discovery Supplier No. K150SHE1
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody calibrator and assay diluent optimized for A 38 detection With a lower limit of detection of 1 95 pg/mL this kit enables the accurate quantification of A 38 for research into Alzheimer s disease and other neurodegenerative disorders