Learn More
Meso Scale Discovery V-PLEX Ab38 Peptide 4G8 Kit
SDP
Catalog No. 5000080573
Encompass
The K150SHE-2 is a thermoelectric module from Mesoscale that can provide cooling or heating It measures 40 x 40 x 3 4mm and provides a maximum heat pumping capacity of 150W The module contains 127 thermocouples and achieves a maximum temperature differential of 70 C It requires a heat sink and fan for proper thermal management The module can be used for temperature control applications including scientific instrumentation medical diagnostics consumer goods and telecommunications equipment With proper system design it provides a compact quiet and reliable solid-state heating and cooling solution
Catalog No. 50-000-80573 Supplier Meso Scale Discovery Supplier No. K150SHE2
The K150SHE-2 is a thermoelectric module from Mesoscale that can provide cooling or heating It measures 40 x 40 x 3 4mm and provides a maximum heat pumping capacity of 150W The module contains 127 thermocouples and achieves a maximum temperature differential of 70 C It requires a heat sink and fan for proper thermal management The module can be used for temperature control applications including scientific instrumentation medical diagnostics consumer goods and telecommunications equipment With proper system design it provides a compact quiet and reliable solid-state heating and cooling solution