Learn More
Meso Scale Discovery V-PLEX Ab38 Peptide 6E10 Kit
SDP
Catalog No. 5000080574
Encompass
The Meso Scale Discovery V-PLEX A 38 Peptide Kit is an electrochemiluminescent immunoassay for measuring A 38 in human CSF and plasma It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The kit includes a 96-well plate with a monoclonal antibody to capture A 38 a detection antibody with an MSD SULFO-TAG label calibrators and necessary buffers and reagents Results are read on MSD s SECTOR Imager instruments and analyzed with MSD Discovery Workbench software The assay quantifies A 38 from 1 57 to 1000 pg/mL
Catalog No. 50-000-80574 Supplier Meso Scale Discovery Supplier No. K150SIE4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX A 38 Peptide Kit is an electrochemiluminescent immunoassay for measuring A 38 in human CSF and plasma It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results The kit includes a 96-well plate with a monoclonal antibody to capture A 38 a detection antibody with an MSD SULFO-TAG label calibrators and necessary buffers and reagents Results are read on MSD s SECTOR Imager instruments and analyzed with MSD Discovery Workbench software The assay quantifies A 38 from 1 57 to 1000 pg/mL