Learn More
Meso Scale Discovery V-PLEX Ab38 Peptide 6E10 Kit
SDP
Catalog No. 5000081601
Encompass
The Meso Scale Discovery V-PLEX A 38 Peptide Kit is an electrochemiluminescent immunoassay for measuring A 38 in human CSF and plasma It uses SULFO-TAG labels and multi-array technology for sensitive results The kit includes a 96-well plate with a monoclonal antibody to A 38 detection antibody with ruthenium calibrators diluent buffers and seals It quantifies from 1 95 to 500 pg/mL requires 50 L of sample and is read on MSD s SECTOR Imager It has high specificity for A 38 with minimal cross-reactivity
Catalog No. 50-000-81601 Supplier Meso Scale Discovery Supplier No. K150SIE2
The Meso Scale Discovery V-PLEX A 38 Peptide Kit is an electrochemiluminescent immunoassay for measuring A 38 in human CSF and plasma It uses SULFO-TAG labels and multi-array technology for sensitive results The kit includes a 96-well plate with a monoclonal antibody to A 38 detection antibody with ruthenium calibrators diluent buffers and seals It quantifies from 1 95 to 500 pg/mL requires 50 L of sample and is read on MSD s SECTOR Imager It has high specificity for A 38 with minimal cross-reactivity