Learn More
Meso Scale Discovery V-PLEX Ab40 Peptide 4G8 Kit
SDP
Catalog No. 5000081603
Encompass
The Meso Scale Discovery V-PLEX A 40 Peptide Kit is designed to quantitatively measure A 40 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity broad dynamic range and low matrix interference The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests It can accurately detect A 40 levels between 12 2-1500 pg/mL with minimal cross-reactivity to A 42 This kit allows reliable measurement of A 40 for research into Alzheimer s disease biomarkers
Catalog No. 50-000-81603 Supplier Meso Scale Discovery Supplier No. K150SJE2
The Meso Scale Discovery V-PLEX A 40 Peptide Kit is designed to quantitatively measure A 40 levels in human samples such as CSF serum and plasma This electrochemiluminescence (ECL) immunoassay utilizes MSD 4G8 antibody-coated plates and SULFO-TAG detection antibodies to provide high sensitivity broad dynamic range and low matrix interference The kit includes a plate detection antibody calibrator diluent and wash buffer for 96 tests It can accurately detect A 40 levels between 12 2-1500 pg/mL with minimal cross-reactivity to A 42 This kit allows reliable measurement of A 40 for research into Alzheimer s disease biomarkers