Learn More
Meso Scale Discovery V-PLEX Ab40 Peptide 4G8 Kit
SDP
Catalog No. 5000081777
Encompass
The Meso Scale Discovery V-PLEX Abeta40 Peptide Kit measures Abeta40 in human serum plasma CSF and cell culture supernatants This ECL-based immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Detection ranges from 1 57 to 1000 pg/mL with a 3 5-hour assay time It supports research on Alzheimer s and neurological disorders involving amyloid beta
Catalog No. 50-000-81777 Supplier Meso Scale Discovery Supplier No. K150SJE1
The Meso Scale Discovery V-PLEX Abeta40 Peptide Kit measures Abeta40 in human serum plasma CSF and cell culture supernatants This ECL-based immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results It includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Detection ranges from 1 57 to 1000 pg/mL with a 3 5-hour assay time It supports research on Alzheimer s and neurological disorders involving amyloid beta