Learn More
Meso Scale Discovery V-PLEX Ab40 Peptide 4G8 Kit
SDP
Catalog No. 5000081854
Encompass
The Meso Scale Discovery V-PLEX A 40 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of A 40 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 96-well 4G8 antibody-coated plate detection antibody standards buffers substrate and plate seals It has a detection range of 50-2500 pg/mL and requires 50 L sample volume per well The assay takes approximately 5 hours to complete This product enables reliable quantification of A 40 levels
Catalog No. 50-000-81854 Supplier Meso Scale Discovery Supplier No. K150SJE4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX A 40 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of A 40 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 96-well 4G8 antibody-coated plate detection antibody standards buffers substrate and plate seals It has a detection range of 50-2500 pg/mL and requires 50 L sample volume per well The assay takes approximately 5 hours to complete This product enables reliable quantification of A 40 levels