Learn More
Meso Scale Discovery V-PLEX Ab40 Peptide 6E10 Kit
SDP
Catalog No. 5000078876
Encompass
The Meso Scale Discovery V-PLEX Abeta40 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Abeta40 in human serum plasma and CSF The kit contains a 96-well plate coated with a monoclonal antibody specific for the N-terminus of Abeta40 a detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 1 6 pg/mL and a detection range of 1 6-1000 pg/mL It can be used to aid in the diagnosis of Alzheimer s disease by measuring Abeta40 levels The kit takes 3 5 hours to run and contains sufficient reagents for one 96-well plate
Catalog No. 50-000-78876 Supplier Meso Scale Discovery Supplier No. K150SKE4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Abeta40 Peptide Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Abeta40 in human serum plasma and CSF The kit contains a 96-well plate coated with a monoclonal antibody specific for the N-terminus of Abeta40 a detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 1 6 pg/mL and a detection range of 1 6-1000 pg/mL It can be used to aid in the diagnosis of Alzheimer s disease by measuring Abeta40 levels The kit takes 3 5 hours to run and contains sufficient reagents for one 96-well plate