Learn More
Meso Scale Discovery V-PLEX Ab42 Peptide 4G8 Kit
SDP
Catalog No. 5000081705
Encompass
The Meso Scale Discovery V-PLEX A 42 Peptide Kit is an electrochemiluminescence immunoassay for the quantitative measurement of A 42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody calibrator and assay diluent optimized to measure A 42 levels between 10-2000 pg/mL This immunoassay allows high-throughput testing of A 42 a peptide associated with Alzheimer s disease and is suitable for research use only
Catalog No. 50-000-81705 Supplier Meso Scale Discovery Supplier No. K150SLE2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX A 42 Peptide Kit is an electrochemiluminescence immunoassay for the quantitative measurement of A 42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a pre-coated plate detection antibody calibrator and assay diluent optimized to measure A 42 levels between 10-2000 pg/mL This immunoassay allows high-throughput testing of A 42 a peptide associated with Alzheimer s disease and is suitable for research use only