Learn More
Meso Scale Discovery V-PLEX Angiogenesis Panel 1 Human Kit, Multiplex Immunoassay, 5 plates, 96-well, 7-spot, detection range: 0.09 - 63,400 pg/mL (varies by analyte), for research use only
SDP
Catalog No. NC2607229
Encompass
The V-PLEX Angiogenesis Panel 1 Human Kit is a highly validated multiplex immunoassay designed for the quantitative determination of seven key human angiogenesis biomarkers. This assay is suitable for research applications in cardiovascular diseases, growth factor studies, and understanding processes like tumor growth and metastatic progression, providing accurate and reproducible results.
  • Assay platform utilizes electrochemiluminescence (ECL) detection
  • Human species reactivity
  • Detects seven analytes including FGF (basic), PlGF, Tie-2, VEGF-A, VEGF-C, VEGF-D, and VEGFR-1/Flt-1
  • 96-well plate type with a 7-spot configuration
  • Includes 5 plates per kit
  • Typical detection ranges from 0.09 to 63,400 pg/mL
  • Compatible with a range of sample types including serum and plasma
  • Research use only; not for diagnostic procedures
  • Compatible instruments include MESO SECTOR and QuickPlex systems
Catalog No. NC2607229 Supplier Meso Scale Discovery Supplier No. K15190D2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Angiogenesis Panel 1 Human Kit is a highly validated multiplex immunoassay designed for the quantitative determination of seven key human angiogenesis biomarkers. This assay is suitable for research applications in cardiovascular diseases, growth factor studies, and understanding processes like tumor growth and metastatic progression, providing accurate and reproducible results.
  • Assay platform utilizes electrochemiluminescence (ECL) detection
  • Human species reactivity
  • Detects seven analytes including FGF (basic), PlGF, Tie-2, VEGF-A, VEGF-C, VEGF-D, and VEGFR-1/Flt-1
  • 96-well plate type with a 7-spot configuration
  • Includes 5 plates per kit
  • Typical detection ranges from 0.09 to 63,400 pg/mL
  • Compatible with a range of sample types including serum and plasma
  • Research use only; not for diagnostic procedures
  • Compatible instruments include MESO SECTOR and QuickPlex systems