Learn More
Meso Scale Discovery V-PLEX GIP active Kit
SDP
Catalog No. 5000080577
Encompass
The Meso Scale Discovery V-PLEX GIP Active Kit is a human GIP immunoassay kit for measuring active GIP in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with a GIP capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve It detects active GIP from 0 15 pg/mL to 1000 pg/mL with high specificity
Catalog No. 50-000-80577 Supplier Meso Scale Discovery Supplier No. K150T5D1
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX GIP Active Kit is a human GIP immunoassay kit for measuring active GIP in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a broad dynamic range and low sample volume requirements The kit includes a 96-well plate with a GIP capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve It detects active GIP from 0 15 pg/mL to 1000 pg/mL with high specificity