Learn More
Meso Scale Discovery V-PLEX GLP-1 Active Kit
SDP
Catalog No. 5000079189
Encompass
The Meso Scale Discovery V-PLEX GLP-1 Active Kit is a sensitive immunoassay for measuring active GLP-1 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with a GLP-1 capture antibody a detection antibody and calibrators for a standard curve It is specific for active GLP-1 with minimal cross-reactivity and offers a rapid 2 5-hour protocol
Catalog No. 50-000-79189 Supplier Meso Scale Discovery Supplier No. K1503OD1
The Meso Scale Discovery V-PLEX GLP-1 Active Kit is a sensitive immunoassay for measuring active GLP-1 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with a GLP-1 capture antibody a detection antibody and calibrators for a standard curve It is specific for active GLP-1 with minimal cross-reactivity and offers a rapid 2 5-hour protocol