Learn More
Meso Scale Discovery V-PLEX GLP-1 Active Kit
SDP
Catalog No. 5000081819
Encompass
The Meso Scale Discovery V-PLEX GLP-1 Active Kit measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with SULFO-TAG calibrators and an assay diluent The active GLP-1 binds to the capture antibody and the detection antibody binds to the analyte An MSD instrument reads the plate applying electricity to emit light quantifying active GLP-1
Catalog No. 50-000-81819 Supplier Meso Scale Discovery Supplier No. K1503OD2
The Meso Scale Discovery V-PLEX GLP-1 Active Kit measures active GLP-1 in serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results It includes a 96-well plate with a GLP-1 capture antibody a detection antibody with SULFO-TAG calibrators and an assay diluent The active GLP-1 binds to the capture antibody and the detection antibody binds to the analyte An MSD instrument reads the plate applying electricity to emit light quantifying active GLP-1