Learn More
Meso Scale Discovery V-PLEX Glucagon Kit 25 PL
SDP
Catalog No. 5000081723
Encompass
The Meso Scale Discovery V-PLEX Glucagon Kit is a highly sensitive immunoassay that quantitatively measures glucagon in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides accurate and reproducible results with a lower limit of detection of 0 7 pg/mL and an upper limit of quantification of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and ancillary reagents needed to complete the assay The total assay time is approximately 3 5 hours This product can aid in the research of pancreatic alpha cell function and diseases like diabetes
Catalog No. 50-000-81723 Supplier Meso Scale Discovery Supplier No. K150U5D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Glucagon Kit is a highly sensitive immunoassay that quantitatively measures glucagon in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides accurate and reproducible results with a lower limit of detection of 0 7 pg/mL and an upper limit of quantification of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and ancillary reagents needed to complete the assay The total assay time is approximately 3 5 hours This product can aid in the research of pancreatic alpha cell function and diseases like diabetes