Learn More
Meso Scale Discovery V-PLEX Glucagon Kit
SDP
Catalog No. 5000081781
Encompass
The Meso Scale Discovery V-PLEX Glucagon Kit is a multiplex immunoassay that quantitatively measures glucagon in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and ancillary reagents needed to run the assay The total assay time is 3 5 hours The kit has a detection range of 1 53-1000 pg/mL and includes reagents for one 96-well plate This assay allows for the accurate quantitation of glucagon from various human samples
Catalog No. 50-000-81781 Supplier Meso Scale Discovery Supplier No. K150U5D2
The Meso Scale Discovery V-PLEX Glucagon Kit is a multiplex immunoassay that quantitatively measures glucagon in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and ancillary reagents needed to run the assay The total assay time is 3 5 hours The kit has a detection range of 1 53-1000 pg/mL and includes reagents for one 96-well plate This assay allows for the accurate quantitation of glucagon from various human samples