Learn More
Meso Scale Discovery V-PLEX Human ASY42 Kit
SDP
Catalog No. 5000078677
Encompass
The Meso Scale Discovery V-PLEX Human A 42 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of amyloid beta 1-42 (A 42) in human cerebrospinal fluid serum and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with a monoclonal antibody specific for the C-terminus of A 42 along with detection antibody calibrator diluent and wash buffer The assay has a typical lower limit of quantification of 1 07 pg/mL and an upper limit of quantification of 700 pg/mL for A 42 It requires 50 L of sample and can perform up to 80 tests in duplicate
Catalog No. 50-000-78677 Supplier Meso Scale Discovery Supplier No. K151LBE4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human A 42 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of amyloid beta 1-42 (A 42) in human cerebrospinal fluid serum and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results The kit includes a 96-well plate coated with a monoclonal antibody specific for the C-terminus of A 42 along with detection antibody calibrator diluent and wash buffer The assay has a typical lower limit of quantification of 1 07 pg/mL and an upper limit of quantification of 700 pg/mL for A 42 It requires 50 L of sample and can perform up to 80 tests in duplicate