Learn More
Meso Scale Discovery V-PLEX Human ASY42 Kit
SDP
Catalog No. 5000079960
Encompass
The V-PLEX Human A 42 Kit from Meso Scale Discovery is an immunoassay for measuring amyloid beta 1-42 (A 42) in human CSF serum and plasma It uses ECL detection technology offering high sensitivity a broad dynamic range and requires only 50 L per well The kit includes a 96-well plate with a monoclonal antibody for the C-terminus of A 42 a detection antibody for the N-terminus conjugated to an ECL label assay diluent calibrator diluent wash buffer read buffer and a calibrator It has high reproducibility and takes about 3 5 hours to run
Catalog No. 50-000-79960 Supplier Meso Scale Discovery Supplier No. K151LBE2
The V-PLEX Human A 42 Kit from Meso Scale Discovery is an immunoassay for measuring amyloid beta 1-42 (A 42) in human CSF serum and plasma It uses ECL detection technology offering high sensitivity a broad dynamic range and requires only 50 L per well The kit includes a 96-well plate with a monoclonal antibody for the C-terminus of A 42 a detection antibody for the N-terminus conjugated to an ECL label assay diluent calibrator diluent wash buffer read buffer and a calibrator It has high reproducibility and takes about 3 5 hours to run