Learn More
Meso Scale Discovery V-PLEX Human C-Peptide Kit
SDP
Catalog No. 5000081739
Encompass
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is a multiplex immunoassay that quantitatively measures C-peptide levels in human serum plasma and culture supernatant samples The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and ancillary reagents required to perform the assay The kit can accurately measure C-peptide levels between 0 008 ng/mL to 2000 ng/mL This product would be useful for researchers studying insulin production and diabetes
Catalog No. 50-000-81739 Supplier Meso Scale Discovery Supplier No. K151X5D1
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is a multiplex immunoassay that quantitatively measures C-peptide levels in human serum plasma and culture supernatant samples The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium calibrators and ancillary reagents required to perform the assay The kit can accurately measure C-peptide levels between 0 008 ng/mL to 2000 ng/mL This product would be useful for researchers studying insulin production and diabetes