Learn More
Meso Scale Discovery V-PLEX Human CRP Kit
SDP
Catalog No. 5000078560
Encompass
The V-PLEX Human CRP Kit from Meso Scale Discovery is an immunoassay that quantitatively measures C-reactive protein (CRP) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The V-PLEX format allows for custom assay development and multiplexing This CRP assay has high sensitivity with a lower limit of detection of 0 02 ng/mL and an upper limit of 10 000 ng/mL The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls to generate a standard curve
Catalog No. 50-000-78560 Supplier Meso Scale Discovery Supplier No. K151STD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human CRP Kit from Meso Scale Discovery is an immunoassay that quantitatively measures C-reactive protein (CRP) in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The V-PLEX format allows for custom assay development and multiplexing This CRP assay has high sensitivity with a lower limit of detection of 0 02 ng/mL and an upper limit of 10 000 ng/mL The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ruthenium and calibrators and controls to generate a standard curve