Learn More
Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit
SDP
Catalog No. 5000079150
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a sensitive immunoassay for measuring human Eotaxin-3 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and a wide dynamic range requiring only 50-100 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an MSD Read Buffer for use with MSD instruments It ensures accurate precise and reproducible quantification of human Eotaxin-3
Catalog No. 50-000-79150 Supplier Meso Scale Discovery Supplier No. K151NUD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a sensitive immunoassay for measuring human Eotaxin-3 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for high sensitivity and a wide dynamic range requiring only 50-100 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and an MSD Read Buffer for use with MSD instruments It ensures accurate precise and reproducible quantification of human Eotaxin-3