Learn More
Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit
SDP
Catalog No. 5000079602
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a sensitive immunoassay for measuring human Eotaxin-3 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology requiring only 50-100 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument like the MESO QuickPlex SQ 120 aiding in the study of chemokine biology and inflammation
Catalog No. 50-000-79602 Supplier Meso Scale Discovery Supplier No. K151NUD1
The Meso Scale Discovery V-PLEX Human Eotaxin-3 Kit is a sensitive immunoassay for measuring human Eotaxin-3 in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology requiring only 50-100 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument like the MESO QuickPlex SQ 120 aiding in the study of chemokine biology and inflammation