Learn More
Meso Scale Discovery V-PLEX Human Eotaxin Kit
SDP
Catalog No. 5000078653
Encompass
The V-PLEX Human Eotaxin Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Eotaxin in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and controls The assay has a sensitivity of 0 06 pg/mL and an upper limit of quantification of 2500 pg/mL This product can help researchers study chemokine biology and inflammation
Catalog No. 50-000-78653 Supplier Meso Scale Discovery Supplier No. K151NSD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Eotaxin Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Eotaxin in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and controls The assay has a sensitivity of 0 06 pg/mL and an upper limit of quantification of 2500 pg/mL This product can help researchers study chemokine biology and inflammation