Learn More
Meso Scale Discovery V-PLEX Human Eotaxin Kit
SDP
Catalog No. 5000081200
Encompass
The Meso Scale Discovery V-PLEX Human Eotaxin Kit is a sensitive immunoassay for measuring human eotaxin in various biological fluids It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and a wide dynamic range requiring 50 L or less per well The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Results are read on MSD s SECTOR Imager providing pg/mL quantitation with an ultra-low detection limit
Catalog No. 50-000-81200 Supplier Meso Scale Discovery Supplier No. K151NSD1
The Meso Scale Discovery V-PLEX Human Eotaxin Kit is a sensitive immunoassay for measuring human eotaxin in various biological fluids It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and a wide dynamic range requiring 50 L or less per well The kit includes a 96-well plate with capture antibody detection antibody with SULFO-TAG calibrators and diluent Results are read on MSD s SECTOR Imager providing pg/mL quantitation with an ultra-low detection limit