Learn More
Meso Scale Discovery V-PLEX Human FGF basic Kit
SDP
Catalog No. 5000079170
Encompass
The V-PLEX Human FGF Basic Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human basic fibroblast growth factor (FGF2) in various biological fluids It includes a 96-well plate with capture antibodies for FGF2 detection antibodies with an electrochemiluminescent label and necessary reagents The assay detects FGF2 in the range of 1 53-1000 pg/mL with minimal cross-reactivity The signal correlates with FGF2 levels in samples
Catalog No. 50-000-79170 Supplier Meso Scale Discovery Supplier No. K151MDD1
The V-PLEX Human FGF Basic Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring human basic fibroblast growth factor (FGF2) in various biological fluids It includes a 96-well plate with capture antibodies for FGF2 detection antibodies with an electrochemiluminescent label and necessary reagents The assay detects FGF2 in the range of 1 53-1000 pg/mL with minimal cross-reactivity The signal correlates with FGF2 levels in samples