Learn More
Meso Scale Discovery V-PLEX Human FGF basic Kit
SDP
Catalog No. 5000079171
Encompass
The V-PLEX Human FGF Basic Kit is a multiplex immunoassay for measuring FGF basic in various biological fluids using ECL technology on the MSD platform It includes a 96-well plate with capture antibodies detection antibody conjugates calibrators controls diluents wash buffer read buffer and plate seals Sensitivity is 1 5 pg/mL with a quantification limit of 10 000 pg/mL It requires 50 L per well and processes up to 80 samples in 3 5 hours For research use only
Catalog No. 50-000-79171 Supplier Meso Scale Discovery Supplier No. K151MDD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human FGF Basic Kit is a multiplex immunoassay for measuring FGF basic in various biological fluids using ECL technology on the MSD platform It includes a 96-well plate with capture antibodies detection antibody conjugates calibrators controls diluents wash buffer read buffer and plate seals Sensitivity is 1 5 pg/mL with a quantification limit of 10 000 pg/mL It requires 50 L per well and processes up to 80 samples in 3 5 hours For research use only