Learn More
Meso Scale Discovery V-PLEX Human FGF basic Kit
SDP
Catalog No. 5000079554
Encompass
The V-PLEX Human FGF Basic Kit from Meso Scale Discovery is a multiplex immunoassay that measures human basic fibroblast growth factor (FGF-2) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The V-PLEX assay requires an MSD instrument such as SECTOR Imager QuickPlex or MESO QuickPlex SQ 120 for analysis The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent
Catalog No. 50-000-79554 Supplier Meso Scale Discovery Supplier No. K151MDD2
The V-PLEX Human FGF Basic Kit from Meso Scale Discovery is a multiplex immunoassay that measures human basic fibroblast growth factor (FGF-2) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The V-PLEX assay requires an MSD instrument such as SECTOR Imager QuickPlex or MESO QuickPlex SQ 120 for analysis The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and sample diluent