Learn More
Meso Scale Discovery V-PLEX Human Flt-1 Kit
SDP
Catalog No. 5000078512
Encompass
The V-PLEX Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human FLT-1 protein in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a detection range of 3 05 - 10 000 pg/mL and requires 25 L of sample The kit has high sensitivity requires no wash steps and results are obtained in as little as 3 5 hours This assay can be used to study angiogenesis cancer and cardiovascular disease involving FLT-1
Catalog No. 50-000-78512 Supplier Meso Scale Discovery Supplier No. K151RZD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay that measures the human FLT-1 protein in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a detection range of 3 05 - 10 000 pg/mL and requires 25 L of sample The kit has high sensitivity requires no wash steps and results are obtained in as little as 3 5 hours This assay can be used to study angiogenesis cancer and cardiovascular disease involving FLT-1