Learn More
Meso Scale Discovery V-PLEX Human Flt-1 Kit
SDP
Catalog No. 5000081295
Encompass
The V-PLEX Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human FLT-1 (VEGFR-1) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on a low background The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a simple reproducible method for FLT-1 measurement in research
Catalog No. 50-000-81295 Supplier Meso Scale Discovery Supplier No. K151RZD2
The V-PLEX Human FLT-1 Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human FLT-1 (VEGFR-1) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection on a low background The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are available in 3 5 hours using an MSD instrument This assay offers a simple reproducible method for FLT-1 measurement in research