Learn More
Meso Scale Discovery V-PLEX Human GM-CSF Kit
SDP
Catalog No. 5000078573
Encompass
The V-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a sensitivity of 0 10 pg/mL and an upper limit of quantification of 1500 pg/mL for GM-CSF The kit can perform up to 80 tests in duplicate and has a testing time of 3 5 hours This highly sensitive and reproducible kit is designed for research use only
Catalog No. 50-000-78573 Supplier Meso Scale Discovery Supplier No. K151RID1
The V-PLEX Human GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of GM-CSF in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a sensitivity of 0 10 pg/mL and an upper limit of quantification of 1500 pg/mL for GM-CSF The kit can perform up to 80 tests in duplicate and has a testing time of 3 5 hours This highly sensitive and reproducible kit is designed for research use only