Learn More
Meso Scale Discovery V-PLEX Human ICAM-1 Kit
SDP
Catalog No. 5000078758
Encompass
The V-PLEX Human ICAM-1 Kit from Meso Scale Discovery is a sandwich immunoassay for measuring human ICAM-1 in serum plasma and cell culture supernatants It includes a 96-well plate with a capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve The assay sensitivity is 0 23 pg/mL with an upper limit of 1500 pg/mL The electrochemiluminescent method offers high sensitivity wide range and small sample volume requirements enabling accurate quantification of human ICAM-1 levels
Catalog No. 50-000-78758 Supplier Meso Scale Discovery Supplier No. K151SUD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human ICAM-1 Kit from Meso Scale Discovery is a sandwich immunoassay for measuring human ICAM-1 in serum plasma and cell culture supernatants It includes a 96-well plate with a capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve The assay sensitivity is 0 23 pg/mL with an upper limit of 1500 pg/mL The electrochemiluminescent method offers high sensitivity wide range and small sample volume requirements enabling accurate quantification of human ICAM-1 levels