Learn More
Meso Scale Discovery V-PLEX Human ICAM-1 Kit
SDP
Catalog No. 5000082223
Encompass
The Meso Scale Discovery V-PLEX Human ICAM-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human ICAM-1 in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human ICAM-1 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 ng/mL and a range from 0 3-2500 ng/mL Sample incubation time is 2 hours The kit can be used to measure soluble ICAM-1 levels as a biomarker of endothelial cell activation and inflammation
Catalog No. 50-000-82223 Supplier Meso Scale Discovery Supplier No. K151SUD2
The Meso Scale Discovery V-PLEX Human ICAM-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human ICAM-1 in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human ICAM-1 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 ng/mL and a range from 0 3-2500 ng/mL Sample incubation time is 2 hours The kit can be used to measure soluble ICAM-1 levels as a biomarker of endothelial cell activation and inflammation