Learn More
Meso Scale Discovery V-PLEX Human IFN-gamma Kit, 25 plates
SDP
Catalog No. NC1218602
Encompass
The Human IFN-gamma Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma in human urine, serum, plasma, and cell culture supernatants.
Catalog No. NC1218602 Supplier Meso Scale Discovery Supplier No. K151QOD4
Add to Cart
Add to Cart
Request a Quote
The Human IFN-gamma Kit provides assay-specific components for the quantitative determination of endogenous IFN-gamma in human urine, serum, plasma, and cell culture supernatants.