Learn More
Meso Scale Discovery V-PLEX Human IL-12p70 Kit
SDP
Catalog No. 5000078478
Encompass
The Meso Scale Discovery V-PLEX Human IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 subunit p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with a monoclonal antibody specific for IL-12p70 a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 1 7-1500 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows for the sensitive and accurate quantification of IL-12p70 levels in under 5 hours
Catalog No. 50-000-78478 Supplier Meso Scale Discovery Supplier No. K151QVD1
The Meso Scale Discovery V-PLEX Human IL-12p70 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 subunit p70 (IL-12p70) in cell culture supernatants serum plasma and other biological fluids This kit contains a 96-well plate coated with a monoclonal antibody specific for IL-12p70 a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 1 7-1500 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows for the sensitive and accurate quantification of IL-12p70 levels in under 5 hours